1,3-Diaminopropane dihydrochloride, Sigma-Aldrich®
Dodavatel: Merck
D23807-100G
D23807-5G
SIALD23807-100GEA
6470
CZK
SIALD23807-100G
SIALD23807-5G
1,3-Diaminopropane dihydrochloride, Sigma-Aldrich®
1,3-Diaminopropane dihydrochloride
Vzorec:
C₃H₁₂Cl₂N₂ |
MDL Number:
MFCD00012525 CAS číslo: 10517-44-9 EINECS: 234-051-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...