Tisk…
Trimesic acid trimethyl ester, Sigma-Aldrich®
Dodavatel: Merck
Synonyms: Trimethyl 1,3,5-benzenetricarboxylate, Trimethyl benzene-1,3,5-tricarboxylate, 1,3,5-Benzenetricarboxylic acid trimethyl ester
M-Clarity™ quality level = MQ100
115983-25G 115983-5G
SIAL115983-25GEA 4770 CZK
SIAL115983-25G SIAL115983-5G
Trimesic acid trimethyl ester, Sigma-Aldrich®
Trimesic acid trimethyl ester
Vzorec: C₁₂H₁₂O₆
MW: 252,22 g/mol
Bod varu: 225…230 °C (0,5 mmHg)
Bod tání: 146,5…147 °C
MDL Number: MFCD00008434
CAS číslo: 2672-58-4
EINECS: 220-215-5