1-(3-Aminopropyl)imidazole 98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
N-(3-Aminopropyl)-imidazole, Imidazole-1-propylamine, 1-(3-Aminopropyl)-1H-imidazole
A14169.18
A14169.30
A14169.18EA
1700
CZK
A14169.18
A14169.30
1-(3-Aminopropyl)imidazole 98%
1-(3-Aminopropyl)imidazole
Purity: 98%
CAS No.: 5036-48-6
Molecular Formula: C6H11N3
Formula Weight: 125.18
Flash Point: 154°(309°F)
Refractive Index: 1.5190
Odour: Amine-like
pH: 11.3
Melting Point: -68°
Boiling Point: 296°
UN No.: 2735
MDL No.: MFCD00009819
Vzorec:
C₆H₁₁N₃ Bod varu: 296 °C Bod tání: –68 °C Hustota: 1,062 Bod vzplanutí: 154 °C (309 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009819 CAS číslo: 5036-48-6 EINECS: 225-730-9 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...