Tisk…

Tyramine hydrochloride, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-(4-Hydroxyphenyl)ethylammonium chloride, 4-(2-Aminoethyl)phenol hydrochloride, 4-Hydroxyphenethylammonium chloride

M-Clarity™ quality level = MQ200

T2879-100G T2879-10MG T2879-1G T2879-25G T2879-5G
SIALT2879-100GEA 15440 CZK
SIALT2879-100G SIALT2879-10MG SIALT2879-1G SIALT2879-25G SIALT2879-5G
Tyramine hydrochloride, Sigma-Aldrich®
Tyramine hydrochloride
Vzorec: C₈H₁₂ClNO
Bod tání: 270…272 °C
MDL Number: MFCD00012901
CAS číslo: 60-19-5
EINECS: 200-462-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR