Tyramine, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ300
W421501-100G
W421501-25G
W421501-SAMPLE
SIALW421501-100GEA
12320
CZK
SIALW421501-100G
SIALW421501-25G
SIALW421501-SAMPLE
Tyramine, Sigma-Aldrich®
Tyramine
Vzorec:
C₈H₁₁NO Bod varu: 325 °C (1013 hPa) Bod tání: 164 °C |
MDL Number:
MFCD00008193 CAS číslo: 51-67-2 EINECS: 200-115-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...