Tyramine, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
T90344-25G
T90344-5G
SIALT90344-25GEA
3910
CZK
SIALT90344-25G
SIALT90344-5G
Tyramine, Sigma-Aldrich®
Tyramine
Vzorec:
C₈H₁₁NO Bod varu: 325 °C (1013 hPa) Bod tání: 164 °C |
MDL Number:
MFCD00008193 CAS číslo: 51-67-2 EINECS: 200-115-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...