Tisk…

(R)-(+)-Limonene 96% unstabilised

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: (R)-(+)-Dipentene, (R)-(+)-1-Methyl-4-(1-methylvinyl)cyclohexene, (R)-(+)-4-Isopropenyl-1-methyl-1-cyclohexene, D(+)-Limonene, (R)-(+)-p-Mentha-1,8-diene, D-Limonene

179390050 179395000 179390025
ACRO179390050EA 960 CZK
ACRO179390050 ACRO179395000 ACRO179390025
(R)-(+)-Limonene 96% unstabilised
(R)-(+)-Limonene
Vzorec: C₁₀H₁₆
MW: 136.24
Bod varu: 175.5
Bod tání: –74 °C
Hustota: 0.8405 g/cm³ (20 °C)
Bod vzplanutí: 48 °C
Storage Temperature: Ambient
MDL Number: MFCD00062991
CAS číslo: 5989-27-5
EINECS: 227-813-5
UN: 2052
ADR: 3 III
Merck Index: 13,05515

Objednat

Výsledky specifikačního testu

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4710 to 1.4740 (20°C, 589 nm)
GC ≥95 %
Relative density (D20/4°C) 0.840 to 0.844
Specific optical rotation +102° to +110° (20°C, 589 nm) (C=1, CH3OH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR