Tisk…

(R)-(-)-Carvone, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

124931-100ML 124931-500ML 124931-5ML
SIAL124931-100MLEA 1500 CZK
SIAL124931-100ML SIAL124931-500ML SIAL124931-5ML
(R)-(-)-Carvone, Sigma-Aldrich®
(R)-(-)-Carvone
Vzorec: C₁₀H₁₄O
Bod varu: 227 °C (1013 hPa)
Hustota: 0,9652 g/cm³ (15 °C)
MDL Number: MFCD00001578
CAS číslo: 6485-40-1
EINECS: 229-352-5

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR