Tisk…

Ethyl (methylthio)acetate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ300

W383503-100G W383503-1KG W383503-SAMPLE
SIALW383503-100GEA 7470 CZK
SIALW383503-100G SIALW383503-1KG SIALW383503-SAMPLE
Ethyl (methylthio)acetate, Sigma-Aldrich®
Ethyl (methylthio)acetate
Vzorec: C₅H₁₀O₂S
MW: 134,2 g/mol
Bod varu: 70 °C (1013 hPa)
Hustota: 1,04 g/cm³ (20 °C)
MDL Number: MFCD00009182
CAS číslo: 4455-13-4
EINECS: 224-700-2
UN: 3272
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR