Tisk…

Bicine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-(Bis(2-hydroxyethyl)amino)acetic acid, N,N-Bis(2-hydroxyethyl)glycine

M-Clarity™ quality level = MQ400

B8660-100G B8660-1KG B8660-250G B8660-25G
SIALB8660-100GEA 11720 CZK
SIALB8660-100G SIALB8660-1KG SIALB8660-250G SIALB8660-25G
Bicine, Sigma-Aldrich®
Bicine
Vzorec: C₆H₁₃NO₄
MW: 163,17 g/mol
Bod tání: 185…192 °C
Storage Temperature: Ambient
MDL Number: MFCD00004295
CAS číslo: 150-25-4
EINECS: 205-755-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR