Tisk…

Bicine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-(Bis(2-hydroxyethyl)amino)acetic acid, N,N-Bis(2-hydroxyethyl)glycine

M-Clarity™ quality level = MQ400

B3876-100G B3876-1KG B3876-250G B3876-25G
SIALB3876-100GEA 5110 CZK
SIALB3876-100G SIALB3876-1KG SIALB3876-250G SIALB3876-25G
Bicine, Sigma-Aldrich®
Bicine
Vzorec: C₆H₁₃NO₄
MW: 163.17
Bod tání: 185…192 °C
Storage Temperature: Ambient
MDL Number: MFCD00004295
CAS číslo: 150-25-4
EINECS: 205-755-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR