(4-Pyridyl)acetic acid ethyl ester ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Ethyl (4-pyridyl)acetate, Ethyl 4-pyridylacetate, Ethyl 4-pyridineacetate
A14550.22
A14550.03
A14550.06
A14550.14
A14550.22EA
37170
CZK
A14550.22
A14550.03
A14550.06
A14550.14
(4-Pyridyl)acetic acid ethyl ester ≥98%
(4-Pyridyl)acetic acid ethyl ester
Purity: 98%
CAS No.: 54401-85-3
Molecular Formula: C9H11NO2
Formula Weight: 165.19
Flash Point:
Vzorec:
C₉H₁₁NO₂ MW: 165,19 g/mol Bod varu: 86…88 °C (1 mmHg) Bod tání: 18…20 °C Hustota: 1,080 Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00051830 CAS číslo: 54401-85-3 EINECS: 259-150-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...