(4-Hydroxyphenyl)acetic acid ethyl ester ≥98%
Dodavatel: Thermo Fisher Scientific
Synonyms:
Ethyl (4-hydroxyphenyl)acetate, Ethyl 2-(4-hydroxyphenyl)acetate
B21832.09
B21832.18
B21832.09EA
2080
CZK
B21832.09
B21832.18
(4-Hydroxyphenyl)acetic acid ethyl ester ≥98%
(4-Hydroxyphenyl)acetic acid ethyl ester
Purity: 98%
CAS No.: 17138-28-2
Molecular Formula: C10H12O3
Formula Weight: 180.21
Melting Point: 36-38°
Boiling Point: 187-189°/20mm
MDL No.: MFCD00016491
Vzorec:
C₁₀H₁₂O₃ MW: 180.2 Bod varu: 187…189 °C (20 mmHg) Bod tání: 36…38 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016491 CAS číslo: 17138-28-2 EINECS: 241-197-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...