(4-Bromophenyl)acetic acid ethyl ester ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Ethyl 2-(4-bromophenyl)acetate, Ethyl (4-bromophenyl)acetate
B22165.14
B22165.06
B22165.22
B22165.14EA
5290
CZK
B22165.14
B22165.06
B22165.22
(4-Bromophenyl)acetic acid ethyl ester ≥98%
(4-Bromophenyl)acetic acid ethyl ester
Purity: 98%
CAS No.: 14062-25-0
Molecular Formula: C10H11BrO2
Formula Weight: 243.10
Melting Point: 29-31°
Boiling Point: 88-90°/0.35mm
MDL No.: MFCD00016333
Vzorec:
C₁₀H₁₁BrO₂ MW: 243.1 Bod varu: 88…90 °C (0.35 mmHg) Bod tání: 29…31 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016333 CAS číslo: 14062-25-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...