(3,5-Dimethoxy-4-hydroxyphenyl)acetic acid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2-(4-Hydroxy-3,5-dimethoxyphenyl)acetic acid, 4-Hydroxy-3,5-dimethoxyphenylacetic acid
M-Clarity™ quality level = MQ100
631310-1G
631310-5G
SIAL631310-1GEA
2890
CZK
SIAL631310-1G
SIAL631310-5G
(3,5-Dimethoxy-4-hydroxyphenyl)acetic acid, Sigma-Aldrich®
(3,5-Dimethoxy-4-hydroxyphenyl)acetic acid
Vzorec:
C₁₀H₁₂O₅ MW: 212,2 g/mol |
MDL Number:
MFCD00016834 CAS číslo: 4385-56-2 EINECS: 224-494-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...