(3,3,3-Trifluoropropyl)trimethoxysilane ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Trimethoxy(3,3,3-trifluoropropyl)silane
L16671.06
L16671.14
L16671.06EA
2440
CZK
L16671.06
L16671.14
(3,3,3-Trifluoropropyl)trimethoxysilane ≥97%
(3,3,3-Trifluoropropyl)trimethoxysilane
Purity: 97%
CAS No.: 429-60-7
Molecular Formula: CF3CH2CH2Si(OCH3)3
Formula Weight: 218.25
Flash Point: 38°(100°F)
Refractive Index: 1.3550
Boiling Point: 144°
Beilstein Reference: 1762962
UN No.: 1993
MDL No.: MFCD00039266
Vzorec:
F₃CCH₂CH₂Si(OCH₃)₃ Bod varu: 144 °C Hustota: 1,140 Bod vzplanutí: 38 °C (100 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00039266 CAS číslo: 429-60-7 EINECS: 207-059-3 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...