(3,3,3-Trifluoropropyl)trimethoxysilane
Dodavatel: abcr
Synonyms:
Trimethoxy(3,3,3-trifluoropropyl)silane
AB111473.0005
AB111473.0025
ABCRAB111473.0005EA
0
CZK
ABCRAB111473.0005
ABCRAB111473.0025
(3,3,3-Trifluoropropyl)trimethoxysilane
(3,3,3-Trifluoropropyl)trimethoxysilane
Vzorec:
F₃CCH₂CH₂Si(OCH₃)₃ Bod varu: 144 °C (1013 hPa) Hustota: 1,137 g/cm³ (20 °C) |
MDL Number:
MFCD00039266 CAS číslo: 429-60-7 EINECS: 207-059-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...