Tisk…

(2-Pyridyl)acetic acid ethyl ester, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Ethyl 2-Pyridylacetate, Ethyl 2-pyridineacetate, Ethyl (2-pyridyl)acetate

E47247-25G E47247-5G
SIALE47247-25GEA 8530 CZK
SIALE47247-25G SIALE47247-5G
(2-Pyridyl)acetic acid ethyl ester, Sigma-Aldrich®
(2-Pyridyl)acetic acid ethyl ester
Vzorec: C₉H₁₁NO₂
MW: 165,19 g/mol
Bod varu: 302 °C (1013 hPa)
Bod tání: 86…90 °C
Hustota: 1,08 g/cm³ (20 °C)
MDL Number: MFCD00006359
CAS číslo: 2739-98-2
EINECS: 220-365-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR