(2-Pyridyl)acetic acid ethyl ester ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Ethyl 2-Pyridylacetate, Ethyl 2-pyridineacetate, Ethyl (2-pyridyl)acetate
B23988.06
B23988.14
B23988.22
B23988.06EA
1860
CZK
B23988.06
B23988.14
B23988.22
(2-Pyridyl)acetic acid ethyl ester ≥98%
(2-Pyridyl)acetic acid ethyl ester
Purity: 98%
CAS No.: 2739-98-2
Molecular Formula: C9H11NO2
Formula Weight: 165.19
Flash Point:
Vzorec:
C₉H₁₁NO₂ MW: 165.19 Bod varu: 69…70 °C (0.05 mmHg) Bod tání: 86…90 °C Hustota: 1.084 Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006359 CAS číslo: 2739-98-2 EINECS: 220-365-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...