Dimethylsulfoxid-D6, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Dimethyl sulphoxide-[²H₆], Hexadeuterodimethyl sulphoxide, DMSO-D6, DMSO-d6, Dimethyl sulphoxide-d6
716731-10ML
716731-50ML
SIAL716731-10MLEA
2200
CZK
SIAL716731-10ML
SIAL716731-50ML
Dimethylsulfoxid-D6, Sigma-Aldrich®
Dimethylsulfoxid-D6
Vzorec:
D₃CS(O)CD₃ MW: 84.09 Bod varu: 189 Bod tání: 20.2 Hustota: 1.19 g/cm³ (20 °C) Bod vzplanutí: 82 °C (ot. pohárek) Storage Temperature: Ambient |
MDL Number:
MFCD00002090 CAS číslo: 2206-27-1 EINECS: 218-617-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...