(2-Bromoethyl)triphenylphosphonium bromide ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
B24833.06
B24833.14
B24833.06EA
1190
CZK
B24833.06
B24833.14
(2-Bromoethyl)triphenylphosphonium bromide ≥98%
(2-Bromoethyl)triphenylphosphonium bromide
Purity: 98%
CAS No.: 7301-93-1
Molecular Formula: BrCH2CH2P(C6H5)3Br
Formula Weight: 450.15
Melting Point: 186-189°
MDL No.: MFCD04039352
Vzorec:
C₂₀H₁₉Br₂P Bod tání: 186…189 °C Storage Temperature: Ambient |
MDL Number:
MFCD04039352 CAS číslo: 7301-93-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...