(1S)-(+)-Camphor-10-sulphonic acid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
((1S,4R)-7,7-Dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulphonic acid, (1S)-(+)-2-Oxobornane-10-sulphonic acid, D(+)-Camphor-10-sulphonic acid, (1S)-(+)-7,7-Dimethylbicyclo(2.2.1)-2-heptanon-1-methane sulphonic acid, (1S)-(+)-10-Camphorsulphonic acid, D-Camphorsulphonic acid
M-Clarity™ quality level = MQ200
C2107-100G
C2107-500G
C2107-5G
SIALC2107-100GEA
1910
CZK
SIALC2107-100G
SIALC2107-500G
SIALC2107-5G
(1S)-(+)-Camphor-10-sulphonic acid, Sigma-Aldrich®
(1S)-(+)-Camphor-10-sulphonic acid
Vzorec:
C₁₀H₁₆O₄S MW: 232.3 Bod varu: 375 Bod tání: 195 |
MDL Number:
MFCD00064157 CAS číslo: 3144-16-9 EINECS: 221-554-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...