Tisk…

(1S)-(+)-Camphor-10-sulphonic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: ((1S,4R)-7,7-Dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulphonic acid, (1S)-(+)-2-Oxobornane-10-sulphonic acid, D(+)-Camphor-10-sulphonic acid, (1S)-(+)-7,7-Dimethylbicyclo(2.2.1)-2-heptanon-1-methane sulphonic acid, (1S)-(+)-10-Camphorsulphonic acid, D-Camphorsulphonic acid

M-Clarity™ quality level = MQ200

21360-100G-F 21360-500G-F
SIAL21360-100G-FEA 1810 CZK
SIAL21360-100G-F SIAL21360-500G-F
(1S)-(+)-Camphor-10-sulphonic acid, Sigma-Aldrich®
(1S)-(+)-Camphor-10-sulphonic acid
Vzorec: C₁₀H₁₆O₄S
MW: 232.3
Bod varu: 375
Bod tání: 195
MDL Number: MFCD00064157
CAS číslo: 3144-16-9
EINECS: 221-554-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR