(1R)-(-)-Camphor-10-sulphonic acid, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
21365-250G-F
21365-50G-F
SIAL21365-250G-FEA
8910
CZK
SIAL21365-250G-F
SIAL21365-50G-F
(1R)-(-)-Camphor-10-sulphonic acid, Sigma-Aldrich®
(1R)-(-)-Camphor-10-sulphonic acid
Vzorec:
C₁₀H₁₆O₄S Bod varu: 375 °C (1013 hPa) Bod tání: 193…196 °C Hustota: 1 g/cm³ (20 °C) |
MDL Number:
MFCD00150753 CAS číslo: 35963-20-3 EINECS: 252-817-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...