Acetonitril ≥99.5% ACS
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
36423.AP
36423.K2
36423.K7
36423.M1
36423.M6
36423.APEA
2240
CZK
36423.AP
36423.K2
36423.K7
36423.M1
36423.M6
Acetonitril ≥99.5% ACS
Acetonitril
Purity: 99.5+%
CAS No.: 75-05-8
Molecular Formula: CH3CN
Formula Weight: 41.05
Flash Point: 5°(41°F)
Physical Form: Liquid
Density: 0.786
Refractive Index: 1.3440
Odour: Pungent
Melting Point: -48° to -45°
Boiling Point: 80-82°
Beilstein Reference: 741857
Merck Reference: 14,70
UN No.: 1648
MDL No.: MFCD00001878
Vzorec:
H₃CCN Bod varu: 80…82 °C Bod tání: –48 °C…–45 °C Hustota: 0,786 Bod vzplanutí: 5 °C (41 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001878 CAS číslo: 75-05-8 EINECS: 200-835-2 UN: 1648 ADR: 3,II REACH: 01-2119471307-38 Merck Index: 14,70 |