Caprinaldehyde 96%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Decylaldehyde, Decanal, Decyl aldehyde
A11656.AE
A11656.AP
A11656.AEEA
1050
CZK
A11656.AE
A11656.AP
Caprinaldehyde 96%
Caprinaldehyde
Purity: 96%
CAS No.: 112-31-2
Molecular Formula: C10H20O
Formula Weight: 156.27
Flash Point: 85°(185°F)
Refractive Index: 1.4280
Melting Point: -5°
Boiling Point: 207-209°
Beilstein Reference: 1362530
MDL No.: MFCD00007031
Vzorec:
C₁₀H₂₀O MW: 156,27 g/mol Bod varu: 207…209 °C Bod tání: –5 °C Hustota: 0,828 Bod vzplanutí: 85 °C (185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007031 CAS číslo: 112-31-2 EINECS: 203-957-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...