Bromoacetaldehyde diethyl acetal 97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
2-Bromo-1,1-diethoxyethane
346521000
346525000
ACRO346521000EA
1330
CZK
ACRO346521000
ACRO346525000
Bromoacetaldehyde diethyl acetal 97%
Bromoacetaldehyde diethyl acetal
Vzorec:
BrH₂CCH(OCH₂CH₃)₂ MW: 197,07 g/mol Bod varu: 66…67 °C (18 mmHg) Hustota: 1,28 g/cm³ Bod vzplanutí: 61 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000214 CAS číslo: 2032-35-1 EINECS: 217-989-1 UN: 2810 ADR: 6.1,II |
Výsledky specifikačního testu
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4380 to 1.4400 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...