1,3-Dibromo-2,2-dimethoxypropane ≥99%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
A11217.14
A11217.06
A11217.22
A11217.14EA
8270
CZK
A11217.14
A11217.06
A11217.22
1,3-Dibromo-2,2-dimethoxypropane ≥99%
1,3-Dibromo-2,2-dimethoxypropane
Purity: 99%
CAS No.: 22094-18-4
Molecular Formula: C5H10Br2O2
Formula Weight: 261.95
Melting Point: 64-67°
Beilstein Reference: 1698992
Vzorec:
BrH₂CC(OCH₃)₂CH₂Br Bod tání: 64…67 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051792 CAS číslo: 22094-18-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...