Tisk…

3-Bromo-1,1,1-trifluoroacetone ≥97%

Dodavatel: Thermo Scientific
Nebezpečí

A14948.22 A14948.06 A14948.14 A14948.PG
A14948.22EA 21830 CZK
A14948.22 A14948.06 A14948.14 A14948.PG
3-Bromo-1,1,1-trifluoroacetone ≥97%
3-Bromo-1,1,1-trifluoroacetone

Purity: 97% CAS No.: 431-35-6 Molecular Formula: BrCH2COCF3 Formula Weight: 190.95 Flash Point: -2°(28°F) Refractive Index: 1.3760 Odour: Pungent Melting Point: -80° Boiling Point: 85-87° Beilstein Reference: 1703387 UN No.: 2924 MDL No.: MFCD00039237

Vzorec: BrH₂CC(O)CF₃
MW: 190,95 g/mol
Bod varu: 85…87 °C
Bod tání: < –80 °C
Hustota: 1,822
Bod vzplanutí: –2 °C (28 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039237
CAS číslo: 431-35-6
EINECS: 207-071-9
UN: 2924
ADR: 3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR