Bromotrichloromethane ≥97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
A10824.22
A10824.36
A10824.22EA
1340
CZK
A10824.22
A10824.36
Bromotrichloromethane ≥97%
Bromotrichloromethane
Purity: 97%
CAS No.: 75-62-7
Molecular Formula: CBrCl3
Formula Weight: 198.28
Flash Point: None
Refractive Index: 1.5060
Odour: Chloroform-like
Melting Point: -6°
Boiling Point: 104-105°
Beilstein Reference: 1732543
UN No.: 2810
MDL No.: MFCD00000783
Vzorec:
BrCl₃C Bod varu: 104…105 °C Bod tání: –6 °C Hustota: 2,015 Storage Temperature: Ambient |
MDL Number:
MFCD00000783 CAS číslo: 75-62-7 EINECS: 200-886-0 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...