3-Bromobutanone ≥97% stabilizovaný
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Stabilization:
Stabiised with magnesium oxide (MgO) < 1 %
A10186.30
A10186.18
A10186.09
A10186.30EA
36310
CZK
A10186.30
A10186.18
A10186.09
3-Bromobutanone ≥97% stabilizovaný
3-Bromobutanone
Stabilized with
Purity: 97%
CAS No.: 814-75-5
Molecular Formula: C4H7BrO
Formula Weight: 151.01
Flash Point:
Vzorec:
H₃CC(O)BrCHCH₃ MW: 151 g/mol Bod varu: 69…70 °C (70 mmHg) Hustota: 1,438 Bod vzplanutí: >100 °C (212 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00013538 CAS číslo: 814-75-5 EINECS: 212-404-6 UN: 2922 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...