Tisk…

1-Bromoktan ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 1-Octyl bromide

A14722.0I A14722.0B A14722.30
A14722.0IEA 16060 CZK
A14722.0I A14722.0B A14722.30
1-Bromoktan ≥98%
1-Bromoktan

Purity: 98+% CAS No.: 111-83-1 Molecular Formula: C8H17Br Formula Weight: 193.13 Flash Point: 78°(172°F) Refractive Index: 1.4518 Odour: Characteristic Melting Point: -55° Boiling Point: 200-202° Beilstein Reference: 1733136 Merck Reference: 14,6764 UN No.: 3082 MDL No.: MFCD00000276

Vzorec: BrH₂C(CH₂)₆CH₃
Bod varu: 200…202 °C
Bod tání: –55 °C
Hustota: 1,114
Bod vzplanutí: 78 °C (172 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000276
CAS číslo: 111-83-1
EINECS: 203-912-9
UN: 3082
ADR: 9,III
Merck Index: 14,6764

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR