1-Bromododecane, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
16970-1L
16970-250ML
SIAL16970-1LEA
3830
CZK
SIAL16970-1L
SIAL16970-250ML
1-Bromododecane, Sigma-Aldrich®
1-Bromododecane
Vzorec:
BrH₂C(CH₂)₁₀CH₃ Bod varu: 276 °C (1013 hPa) Bod tání: –10 °C Hustota: 1,04 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000225 CAS číslo: 143-15-7 EINECS: 205-587-9 Merck Index: 13,05406 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...