1-Bromodecane ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
1-Decyl bromide
A11340.0C
A11340.0E
A11340.22
A11340.36
A11340.0CEA
24950
CZK
A11340.0C
A11340.0E
A11340.22
A11340.36
1-Bromodecane ≥98%
1-Bromodecane
Purity: 98%
CAS No.: 112-29-8
Molecular Formula: C10H21Br
Formula Weight: 221.19
Flash Point: 94°(201°F)
Refractive Index: 1.4560
Melting Point: -30°
Boiling Point: 238°
Beilstein Reference: 1735227
UN No.: 2810
MDL No.: MFCD00000221
Vzorec:
BrH₂C(CH₂)₈CH₃ MW: 221,18 g/mol Bod varu: 238 °C Bod tání: –30 °C Hustota: 1,067 Bod vzplanutí: 94 °C (201 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000221 CAS číslo: 112-29-8 EINECS: 203-955-3 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...