1-Brombutan ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
A10696.0C
A10696.0E
A10696.36
A10696.0CEA
12940
CZK
A10696.0C
A10696.0E
A10696.36
1-Brombutan ≥98%
1-Bromobutane
Purity: 98+%
CAS No.: 109-65-9
Molecular Formula: C4H9Br
Formula Weight: 137.03
Flash Point: 20°(68°F)
Physical Form: Liquid
Density: 1.275
Refractive Index: 1.4400
Appearance: Colourless
Odour: Characteristic
Melting Point: -112°
Boiling Point: 101-102°
Beilstein Reference: 1098260
Merck Reference: 14,1553
UN No.: 1126
MDL No.: MFCD00000260
Vzorec:
BrH₂C(CH₂)₂CH₃ Bod varu: 101…102 °C Bod tání: –112 °C Hustota: 1,275 Bod vzplanutí: 20 °C (68 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000260 CAS číslo: 109-65-9 EINECS: 203-691-9 UN: 1126 ADR: 3,II Merck Index: 14,1553 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...