Tisk…

1,8-Dibromooctane ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: Octamethylendibromide

A15651.14 A15651.18 A15651.22 A15651.30
A15651.14EA 1680 CZK
A15651.14 A15651.18 A15651.22 A15651.30
1,8-Dibromooctane ≥98%
1,8-Dibromooctane

Purity: 98% CAS No.: 4549-32-0 Molecular Formula: Br(CH2)8Br Formula Weight: 272.02 Flash Point: 110°(230°F) Refractive Index: 1.4995 Melting Point: 15-16° Boiling Point: 270-272° Beilstein Reference: 1698114 MDL No.: MFCD00000277

Vzorec: BrH₂C(CH₂)₆CH₂Br
MW: 272,02 g/mol
Bod varu: 270…272 °C
Bod tání: 15…16 °C
Hustota: 1,468
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000277
CAS číslo: 4549-32-0
EINECS: 224-912-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR