Tisk…

1,6-Dibromohexane ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Hexamethylen dibromide

A13417.22 A13417.36
A13417.22EA 930 CZK
A13417.22 A13417.36
1,6-Dibromohexane ≥97%
1,6-Dibromohexane

Purity: 97+% CAS No.: 629-03-8 Molecular Formula: Br(CH2)6Br Formula Weight: 243.98 Flash Point: 110°(230°F) Refractive Index: 1.5075 Odour: Odourless Melting Point: -2° Boiling Point: 242-244° Beilstein Reference: 1236322 UN No.: 3082 MDL No.: MFCD00000272

Vzorec: BrH₂C(CH₂)₄CH₂Br
Bod varu: 242…244 °C
Bod tání: –2 °C
Hustota: 1,610
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000272
CAS číslo: 629-03-8
EINECS: 211-067-2
UN: 3082
ADR: 9,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR