Tisk…

2-Bromopropene ≥99% stabilizovaný

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 2-Propenyl bromide, α-Methylvinyl bromide, Isopropenyl bromide

Stabilization: Stabilised with copper

L14229.06 L14229.14 L14229.22
L14229.06EA 1600 CZK
L14229.06 L14229.14 L14229.22
2-Bromopropene ≥99% stabilizovaný
2-Bromopropene

Stabilized


Purity: 99% CAS No.: 557-93-7 Molecular Formula: C3H5Br Formula Weight: 120.98 Storage Temperature: 2°C to 8°C Flash Point: 4°(39°F) Refractive Index: 1.4430 Odour: Pungent Melting Point: -87° Boiling Point: 47-49° Beilstein Reference: 1731926 UN No.: 1993 MDL No.: MFCD00000140

Vzorec: C₃H₅Br
MW: 120,98 g/mol
Bod varu: 47…49 °C
Bod tání: –87 °C
Hustota: 1,362
Bod vzplanutí: 4 °C (39 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00000140
CAS číslo: 557-93-7
EINECS: 209-185-4
UN: 1993
ADR: 3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR