Tisk…

2-Iodo-2-methylpropane 95% stabilised, powder

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: tert-Butyl iodide, 2-Methyl-2-propyliodide

Stabilization: Stabilised with copper

L02786.14 L02786.22
L02786.14EA 2070 CZK
L02786.14 L02786.22
2-Iodo-2-methylpropane 95% stabilised, powder
2-Iodo-2-methylpropane

Purity: 95% CAS No.: 558-17-8 Molecular Formula: C4H9I Formula Weight: 184.02 Flash Point: 7°(44°F) Refractive Index: 1.4910 Odour: Recognizable Melting Point: -38° Boiling Point: 99-100° Beilstein Reference: 505950 UN No.: 2391 MDL No.: MFCD00001068

Vzorec: C₄H₉I
Bod varu: 99…100 °C
Bod tání: –38 °C
Hustota: 1,547
Bod vzplanutí: 7 °C (44 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001068
CAS číslo: 558-17-8
EINECS: 209-190-1
UN: 2391
ADR: 3,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR