2-Iodobutane ≥99% stabilizovaný
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Stabilization:
Stabilised with copper
B20619.18
B20619.30
B20619.36
B20619.18EA
1230
CZK
B20619.18
B20619.30
B20619.36
2-Iodobutane ≥99% stabilizovaný
2-Iodobutane
Stabilized with copper
Purity: 99%
CAS No.: 513-48-4
Molecular Formula: C4H9I
Formula Weight: 184.02
Flash Point: 21°(70°F)
Refractive Index: 1.4991
Odour: Ether-like
Melting Point: -104°
Boiling Point: 119-120°
Beilstein Reference: 1718777
Merck Reference: 14,1574
UN No.: 2390
MDL No.: MFCD00001072
Vzorec:
C₄H₉I Bod varu: 119…120 °C Bod tání: –104 °C Hustota: 1,597 Bod vzplanutí: 21 °C (70 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001072 CAS číslo: 513-48-4 EINECS: 208-163-1 UN: 2390 ADR: 3,II Merck Index: 14,1574 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...