Calcium nitrate hydrate ≥99.9995% (metals basis), Puratronic®
Dodavatel: Thermo Scientific
|
Nebezpečí
|
44515.06
44515.14
44515.22
44515.06EA
1700
CZK
44515.06
44515.14
44515.22
Calcium nitrate hydrate ≥99.9995% (metals basis), Puratronic®
Calcium nitrate hydrate
Purity: 99.9995%
CAS No.: 35054-52-5
Molecular Formula: CaN2O6.xH2O
Formula Weight: 164.09(anhy)
Physical Form: Crystalline
Density: 1.82
Appearance: White
Melting Point: 42°
Boiling Point: 132° dec.
Merck Reference: 14,1682
UN No.: 1454
MDL No.: MFCD00149604
Vzorec:
Ca(NO₃)₂·H₂O MW: 182,1 g/mol Bod varu: 132 °C (rozkl.) Bod tání: 42 °C Hustota: 1,82 Storage Temperature: Ambient |
MDL Number:
MFCD00149604 CAS číslo: 35054-52-5 EINECS: 233-332-1 UN: 1454 ADR: 5.1,III Merck Index: 14,1682 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...