Calcium chloride hydrate ≥99.9965% (metals basis), Puratronic®
Dodavatel: Thermo Scientific
|
Varování
|
10680.04
10680.09
10680.18
10680.30
10680.04EA
1670
CZK
10680.04
10680.09
10680.18
10680.30
Calcium chloride hydrate ≥99.9965% (metals basis), Puratronic®
Calcium chloride hydrate
Purity: 99.9965%
CAS No.: 7774-34-7
Molecular Formula: CaCl2.xH2O (x≈4-6)
Formula Weight: 110.99(anhy)
Physical Form: Crystalline Aggregates
Density: 1.71
Merck Reference: 14,1659
MDL No.: MFCD00149616
Vzorec:
CaCl₂·H₂O MW: 110,98 g/mol (anhy) Bod tání: 29,54 °C Hustota: 1,71 Storage Temperature: Ambient |
MDL Number:
MFCD00149613 CAS číslo: 22691-02-7 EINECS: 607-129-7 Merck Index: 14,1659 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...