Tisk…

Sodium thiophosphate hydrate

Dodavatel: Thermo Scientific
Varování

56146.06 56146.14
56146.06EA 2740 CZK
56146.06 56146.14
Sodium thiophosphate hydrate
Sodium thiophosphate hydrate

CAS No.: 10489-48-2 Molecular Formula: Na3PO3S.xH2O Formula Weight: 179.93(anhy) Physical Form: Crystalline aggregates Appearance: White to green Melting Point: 53-58° MDL No.: MFCD00041513

Vzorec: H₂Na₃O₄PS
MW: 198,015 g/mol
Bod tání: 53…58 °C
Hustota: 1,67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00041513
CAS číslo: 10489-48-2
EINECS: 231-867-5
Merck Index: 14,8693

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR