Dihydrogenfosforečnan sodný dihydrát, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Phosphoric acid (sodium hydrate), Sodium phosphate monobasic dihydrate, Sodium dihydrogen orthophosphate dihydrate
71500-250G
71500-1KG
71500-5KG
SIAL71500-250GEA
929
CZK
SIAL71500-250G
SIAL71500-1KG
SIAL71500-5KG
Dihydrogenfosforečnan sodný dihydrát, Sigma-Aldrich®
Dihydrogenfosforečnan sodný dihydrát
Vzorec:
NaH₂PO₄·2H₂O MW: 156,01 g/mol Bod varu: ∼650 °C (1013 hPa) Bod tání: 60 °C Hustota: 1,91 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149209 CAS číslo: 13472-35-0 EINECS: 231-449-2 REACH: 01-2119489796-13 Merck Index: 12,08806 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...