Hydrogenfosforečnan dvojsodný dihydrát, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
sec-Sodium phosphate, Sodium monohydrogen phosphate dihydrate, Sodium hydrogenphosphate dihydrate, di-Sodium hydrogen orthophosphate dihydrate, Sodium phosphate dibasic dihydrate, Disodium phosphate, di-Sodium hydrogen phosphate dihydrate, Sorensen's salt
M-Clarity™ quality level = MQ300
71645-1KG
71645-5KG
SIAL71645-1KGEA
2670
CZK
SIAL71645-1KG
SIAL71645-5KG
Hydrogenfosforečnan dvojsodný dihydrát, Sigma-Aldrich®
Hydrogenfosforečnan dvojsodný dihydrát
Vzorec:
Na₂HPO₄·2H₂O Hustota: 2,1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149182 CAS číslo: 10028-24-7 EINECS: 231-448-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...