Dichroman sodný dihydrát 98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
L14890.22
L14890.36
L14890.PG
L14890.22EA
800
CZK
L14890.22
L14890.36
L14890.PG
Dichroman sodný dihydrát 98%
Sodium dichromate dihydrate
Purity: 99%
CAS No.: 7789-12-0
Molecular Formula: Cr2Na2O7.2H2O
Formula Weight: 298.00 (261.97anhy)
Odour: Odourless
Melting Point: 100° -2H2O
UN No.: 3087
MDL No.: MFCD00149166
Vzorec:
Na₂Cr₂O₇·2H₂O MW: 298,00 g/mol (261,97 anhy) Bod tání: 100 °C (-2H₂O) Hustota: 2,348 Storage Temperature: Ambient |
MDL Number:
MFCD00149166 CAS číslo: 7789-12-0 EINECS: 234-190-3 UN: 3087 ADR: 5.1,II Merck Index: 14,8609 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...