Tisk…

(R)-(+)-Limonene, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: (R)-(+)-Dipentene, (R)-(+)-1-Methyl-4-(1-methylvinyl)cyclohexene, (R)-(+)-4-Isopropenyl-1-methyl-1-cyclohexene, D(+)-Limonene, (R)-(+)-p-Mentha-1,8-diene, D-Limonene

M-Clarity™ quality level = MQ100

62122-1L-F 62122-250ML-F
SIAL62122-1L-FEA 3830 CZK
SIAL62122-1L-F SIAL62122-250ML-F
(R)-(+)-Limonene, Sigma-Aldrich®
(R)-(+)-Limonene
Vzorec: C₁₀H₁₆
MW: 136.24
Bod varu: 175.5
Bod tání: –74 °C
Hustota: 0.8411 g/cm³ (20 °C)
Bod vzplanutí: 43
Storage Temperature: Ambient
MDL Number: MFCD00062991
CAS číslo: 5989-27-5
EINECS: 227-813-5
UN: 2052
ADR: 3 III
Merck Index: 13,05515

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR