(R)-(+)-Limonene 97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
(R)-(+)-Dipentene, (R)-(+)-1-Methyl-4-(1-methylvinyl)cyclohexene, (R)-(+)-4-Isopropenyl-1-methyl-1-cyclohexene, D(+)-Limonene, (R)-(+)-p-Mentha-1,8-diene, D-Limonene
Stabilized
Purity: 97%
CAS No.: 5989-27-5
Molecular Formula: C10H16
Formula Weight: 136.23
Flash Point: 48°(118°F)
Refractive Index: 1.4730
Odour: Fruit-like
Melting Point: -74°
Boiling Point: 177-178°
Beilstein Reference: 2204754
Merck Reference: 14,5493
Optical Rotation: +115° (c=10 in ethanol)
UN No.: 2052
MDL No.: MFCD00062991
Vzorec:
C₁₀H₁₆ MW: 136.24 Bod varu: 177…178 °C Bod tání: –74 °C Hustota: 0.841 Bod vzplanutí: 48 °C (118 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00062991 CAS číslo: 5989-27-5 EINECS: 227-813-5 UN: 2052 ADR: 3 III Merck Index: 13,05515 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...