(S)-(+)-sec-Butylamine 98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
(S)-(+)-2-Aminobutane, (S)-(+)-2-Butylamine
L10069.03
L10069.06
L10069.MD
L10069.03EA
5630
CZK
L10069.03
L10069.06
L10069.MD
(S)-(+)-sec-Butylamine 98%
(S)-(+)-sec-Butylamine
Purity: 98%
CAS No.: 513-49-5
Molecular Formula: C4H11N
Formula Weight: 73.14
Flash Point: -19°(-2°F)
Refractive Index: 1.3930
Odour: Amine-like
pH: 12.5
Boiling Point: 63°
Beilstein Reference: 1718760
Merck Reference: 14,1544
Optical Rotation: +7.5° (neat)
UN No.: 2733
MDL No.: MFCD00064417
Vzorec:
C₄H₁₁N Bod varu: 63 °C Bod tání: –104 °C Hustota: 0,722 Bod vzplanutí: –19 °C (–2 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00064417 CAS číslo: 513-49-5 EINECS: 208-164-7 UN: 2733 ADR: 3,II Merck Index: 14,1544 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...