Tisk…

(R)-(-)-sec-Butylamine 99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: (R)-(-)-2-Butylamine, (R)-(-)-2-Aminobutane

L03889.03 L03889.06 L03889.MD
L03889.03EA 6320 CZK
L03889.03 L03889.06 L03889.MD
(R)-(-)-sec-Butylamine 99%
(R)-(-)-sec-Butylamine

Purity: 99% CAS No.: 13250-12-9 Molecular Formula: C4H11N Formula Weight: 73.14 Flash Point: -19°(-2°F) Refractive Index: 1.3930 Odour: Amine-like pH: 12.3 Boiling Point: 63° Beilstein Reference: 1718760 Merck Reference: 14,1544 Optical Rotation: -7.5° (neat) UN No.: 2733 MDL No.: MFCD00064416

Vzorec: C₄H₁₁N
Bod varu: 63 °C
Bod tání: –104 °C
Hustota: 0,722
Bod vzplanutí: –19 °C (–2 °F)
Storage Temperature: Ambient
MDL Number: MFCD00064416
CAS číslo: 13250-12-9
EINECS: 236-232-6
UN: 2733
ADR: 3,II
Merck Index: 14,1544

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR